4-Iodo-N,N-dimethylaniline 98%

Supplier: Ambeed
Warning

A489054-100MG A489054-10G A489054-250MG A489054-1G A489054-5G
103828-200EA 13 USD
103828-200 103839-272 76830-034 76830-036 76830-038 76843-226 76843-228
4-Iodo-N,N-dimethylaniline 98%
4-Iodo-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀IN
MW: 247.0800 g/mol
Boiling Pt: 263.7 °C (760 mm Hg)
Flash Pt: 113.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00457548
CAS Number: 698-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR