4-Iodoisoxazole 98%

Supplier: Ambeed
Warning

A108335-250MG A108335-1G
103797-328EA 25 USD
103797-328 103823-110 76780-958 77309-956
4-Iodoisoxazole 98%
4-Iodoisoxazole

Precaución: for Research Use Only

Formula: C₃H₂INO
MW: 194.9600 g/mol
Boiling Pt: 214.8 °C (760 mm Hg)
Flash Pt: 83.724 °C
Storage Temperature: Freezer
MDL Number: MFCD09999222
CAS Number: 847490-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR