α,α,α-Trifluoro-4-iodotoluene ≥98% stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilized with Copper chip
A161839-100G
A161839-10G
A161839-500G
A161839-5G
A161839-25G
A161839-1000G
103843-786EA
5
USD
103843-786
76768-580
76768-582
76768-584
77300-562
77319-800
77567-364
α,α,α-Trifluoro-4-iodotoluene ≥98% stabilized
α,α,α-Trifluoro-4-iodotoluene
Precaución: For research use only.
Formula:
C₇H₄F₃I Boiling Pt: 186.3 °C (760 mm Hg) Melting Pt: …8.3 °C Flash Pt: 69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039398 CAS Number: 455-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...