4-Iodobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A211346-100G
A211346-10G
A211346-500G
A211346-1G
A211346-25G
A211346-5G
103818-096EA
9
USD
103818-096
103848-790
77139-600
77139-602
77139-604
77303-182
77303-184
77303-186
4-Iodobenzaldehyde 98%
4-Iodobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅IO MW: 232.02 g/mol Boiling Pt: 264.8 °C (760 mm Hg) Melting Pt: 78…82 °C Flash Pt: 114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039576 CAS Number: 15164-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...