4-Iodoanisole 98%

Supplier: Ambeed
Warning

A111398-100G A111398-10G A111398-1G A111398-25G A111398-500G A111398-5G
103823-644EA 6 USD
103823-644 77107-964 77107-966 77107-968 77107-970 77107-972 77284-106
4-Iodoanisole 98%
4-Iodoanisole

Precaución: for Research Use Only

Formula: C₇H₇IO
Boiling Pt: 239 °C (760 mm Hg)
Melting Pt: 46…51 °C
Flash Pt: 95.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00001056
CAS Number: 696-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR