4-Iodo-6-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A160167-250MG A160167-1G
103805-342EA 77 USD
103805-342 103843-572 77011-176 77300-444
4-Iodo-6-methoxypyrimidine 97%
4-Iodo-6-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅IN₂O
MW: 236.0100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07787396
CAS Number: 161489-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR