4-Iodo-3-methylphenol 96%

Supplier: Ambeed
Warning

A127680-10G A127680-1G A127680-5G
103801-000EA 20 USD
103801-000 77285-164 77313-258 77313-260
4-Iodo-3-methylphenol 96%
4-Iodo-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₇IO
MW: 234.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09029708
CAS Number: 133921-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR