4-Iodo-3,5-xylenol 97%
Supplier: Ambeed
|
Warning
|
A238015-100G
A238015-10G
A238015-250MG
A238015-1G
A238015-25G
A238015-5G
103811-194EA
15
USD
103811-194
103850-978
76692-206
76692-208
76692-210
77090-074
77090-076
77090-078
4-Iodo-3,5-xylenol 97%
4-Iodo-3,5-xylenol
Precaución: for Research Use Only
Formula:
C₈H₉IO Boiling Pt: 293 °C (760 mm Hg) Melting Pt: 129…133 °C Flash Pt: 131 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00174284 CAS Number: 80826-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...