4-Iodo-2-nitroaniline 97%

Supplier: Ambeed
Warning

A718885-5G A718885-25G A718885-1G A718885-10G
103840-726EA 14 USD
103840-726 76844-942 77118-036 77308-190 77686-708
4-Iodo-2-nitroaniline 97%
4-Iodo-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅IN₂O₂
Boiling Pt: 352.9 °C (760 mm Hg)
Melting Pt: 120…123 °C
Flash Pt: 167.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06808526
CAS Number: 20691-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR