4-Iodo-2-methylpyridine 98%

Supplier: Ambeed
Warning

A195653-100MG A195653-250MG
103816-770EA 98 USD
103816-770 103847-382 77287-540 77287-542
4-Iodo-2-methylpyridine 98%
4-Iodo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆IN
MW: 219.023 g/mol
Boiling Pt: 222.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09702484
CAS Number: 22282-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR