4-Iodo-2,6-dimethylaniline 97%
Supplier: Ambeed
|
Warning
|
A111512-250MG
A111512-1G
A111512-10G
103797-852EA
25
USD
103797-852
103823-672
76970-820
77310-478
77570-652
4-Iodo-2,6-dimethylaniline 97%
4-Iodo-2,6-dimethylaniline
Precaución: for Research Use Only
Formula:
C₈H₁₀IN Boiling Pt: 290.6 °C (760 mm Hg) Melting Pt: 50…53 °C Flash Pt: 129.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08061611 CAS Number: 4102-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...