4-Iodo-2-(trifluoromethyl)pyridine 97%
Supplier: Ambeed
|
Warning
|
A203381-1G
A203381-5G
A203381-10G
A203381-250MG
103817-458EA
38
USD
103817-458
103848-112
77086-248
77086-250
77112-256
77112-258
4-Iodo-2-(trifluoromethyl)pyridine 97%
4-Iodo-2-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₃F₃IN Boiling Pt: 206 °C (760 mm Hg) Melting Pt: 24…26 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08234939 CAS Number: 590371-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...