4-Iodo-2-(trifluoromethyl)benzonitrile 97%
Supplier: Ambeed
|
Warning
|
A750995-1G
A750995-25G
A750995-5G
A750995-10G
A750995-250MG
103840-848EA
15
USD
103840-848
76845-080
76845-082
76845-084
77293-518
77293-520
4-Iodo-2-(trifluoromethyl)benzonitrile 97%
4-Iodo-2-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₃F₃IN Boiling Pt: 282.3 °C (760 mm Hg) Melting Pt: 50…55 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458091 CAS Number: 101066-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...