4-Iodo-1-tritylimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-1-trityl-1H-imidazole

A213135-250MG A213135-1G A213135-5G A213135-10G
103818-264EA 6 USD
103818-264 103848-966 76721-776 77087-446 77087-448 77600-720
4-Iodo-1-tritylimidazole 97%
4-Iodo-1-tritylimidazole

Precaución: for Research Use Only

Formula: C₂₂H₁₇IN₂
MW: 436.2900 g/mol
Boiling Pt: 509.7 °C (760 mm Hg)
Melting Pt: 223 °C
Flash Pt: 262 °C
Storage Temperature: Ambient
MDL Number: MFCD02179542
CAS Number: 96797-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR