4-Iodo-1-methylpyridin-2(1H)-one 97%
Supplier: Ambeed
|
Warning
|
A565035-100MG
A565035-250MG
A565035-1G
A565035-5G
103828-768EA
88
USD
103828-768
103839-906
77017-758
77017-760
77307-822
77307-824
4-Iodo-1-methylpyridin-2(1H)-one 97%
4-Iodo-1-methylpyridin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₆H₆INO MW: 235.02 g/mol Boiling Pt: 276.9±29.0 °C (760 mm Hg) Flash Pt: 121.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD15474993 CAS Number: 889865-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...