4-Iodo-1-methylpyrazole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-1-methyl-1H-pyrazole-5-carboxylic acid

A207088-250MG A207088-1G A207088-5G
103817-760EA 22 USD
103817-760 103848-440 77030-324 77086-710 77086-712
4-Iodo-1-methylpyrazole-5-carboxylic acid 98%
4-Iodo-1-methylpyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅IN₂O₂
MW: 252.01 g/mol
Boiling Pt: 362.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00461121
CAS Number: 75092-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR