4-Iodo-2-methylanisole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-1-methoxy-2-methylbenzene

A123243-100G A123243-10G A123243-25G A123243-1G A123243-5G
103800-370EA 7 USD
103800-370 77246-192 77246-194 77246-196 77297-936 77312-486
4-Iodo-2-methylanisole 98%
4-Iodo-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉IO
MW: 248.06 g/mol
Boiling Pt: 247.7 °C (760 mm Hg)
Melting Pt: 75…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425972
CAS Number: 75581-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR