4-Iodobiphenyl 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Iodo-1,1'-biphenyl
A124452-100G
A124452-10G
A124452-25G
A124452-500G
A124452-5G
76996-012EA
39
USD
76996-012
76996-014
76996-016
76996-018
77312-670
4-Iodobiphenyl 97%
4-Iodobiphenyl
Precaución: for Research Use Only
Formula:
C₁₂H₉I Boiling Pt: 322.7 °C (760 mm Hg) Melting Pt: 110…114 °C Flash Pt: 145.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019028 CAS Number: 1591-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...