4-Indanol 97%

Supplier: Ambeed
Warning

A123840-100G A123840-10G A123840-250MG A123840-1G A123840-5G
103800-448EA 13 USD
103800-448 77285-008 77285-010 77285-012 77312-566 77312-568
4-Indanol 97%
4-Indanol

Precaución: for Research Use Only

Formula: C₉H₁₀O
Storage Temperature: Refrigerator
MDL Number: MFCD00053431
CAS Number: 1641-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR