4-Imidazol-1-yl-phenylamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(1H-Imidazol-1-yl)aniline
A161675-10G
A161675-250MG
A161675-1G
A161675-25G
A161675-5G
103805-514EA
15
USD
103805-514
103843-752
76997-810
76997-812
77319-740
77319-742
77319-744
4-Imidazol-1-yl-phenylamine 97%
4-Imidazol-1-yl-phenylamine
Precaución: for Research Use Only
Formula:
C₉H₉N₃ MW: 159.1900 g/mol Boiling Pt: 355.4 °C (760 mm Hg) Melting Pt: 143…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074865 CAS Number: 2221-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...