4-Imidazol-1-ylmethylphenylamine 98%

Supplier: Ambeed
Warning

Synonyms: 4-[(1H-Imidazol-1-yl)methyl]aniline

A176992-1G A176992-25G A176992-250MG A176992-5G
103807-194EA 45 USD
103807-194 77082-892 77082-894 77287-026 77287-028
4-Imidazol-1-ylmethylphenylamine 98%
4-Imidazol-1-ylmethylphenylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃
MW: 173.2144 g/mol
Boiling Pt: 387.6 °C (760 mm Hg)
Melting Pt: 128…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06660785
CAS Number: 56643-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR