4-Imidazol-1-ylbenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(1-Imidazolyl)benzaldehyde, 1-(4-Formylphenyl)imidazole, 4-(1H-Imidazol-1-yl)benzaldehyde
A211387-100G
A211387-10G
A211387-25G
A211387-5G
A211387-1G
103818-104EA
6
USD
103818-104
103848-798
76800-646
76800-648
77087-202
77087-204
77303-194
4-Imidazol-1-ylbenzaldehyde 98%
4-Imidazol-1-ylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O MW: 172.1833 g/mol Melting Pt: 153…155 °C Flash Pt: 200 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00209977 CAS Number: 10040-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...