Tetrahydro-2H-thiopyran-4-ol-1,1-dioxide 98%
Supplier: Ambeed
|
Warning
|
A102675-100MG
A102675-250MG
A102675-1G
103796-406EA
61
USD
103796-406
103822-104
76994-582
76994-584
77308-972
Tetrahydro-2H-thiopyran-4-ol-1,1-dioxide 98%
Tetrahydro-2H-thiopyran-4-ol-1,1-dioxide
Precaución: for Research Use Only
Formula:
C₅H₁₀O₃S Storage Temperature: Ambient |
MDL Number:
MFCD12152579 CAS Number: 194152-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...