4-Hydroxyquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A101550-100G A101550-10G A101550-25G A101550-500G A101550-5G
103821-706EA 12 USD
103821-706 76809-258 76809-260 76809-262 76809-264 77297-148
4-Hydroxyquinolin-2(1H)-one 97%
4-Hydroxyquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: >300 °C
Flash Pt: 137.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00277932
CAS Number: 86-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR