4-Hydroxypyridine-2,6-dicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Helidaminic acid, 2,6-Dicarboxy-4-hydroxypyridine, 4-Hydroxydipicolinic acid

A322684-100G A322684-10G A322684-1G A322684-25G A322684-5G
103825-308EA 10 USD
103825-308 103836-242 77141-846 77141-848 77306-266 77306-268 77306-270
4-Hydroxypyridine-2,6-dicarboxylic acid 98%
4-Hydroxypyridine-2,6-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MW: 183.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066478
CAS Number: 499-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR