4-Hydroxypicolinic acid, (max. 10% H₂O) 95% (contains < 10% H₂O)

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxypicolinic acid, 4-Hydroxy-2-pyridinecarboxylic acid, 4-Hydroxypyridine-2-carboxylic acid

A142215-100G A142215-100MG A142215-10G A142215-250MG A142215-1G
103803-126EA 9 USD
103803-126 77285-800 77285-802 77285-804 77285-806 77316-340
4-Hydroxypicolinic acid, (max. 10% H₂O) 95% (contains < 10% H₂O)
4-Hydroxypicolinic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 313.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00521155
CAS Number: 22468-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR