4-Hydroxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

A137634-1000G A137634-100G A137634-10G A137634-500G A137634-25G A137634-5G A137634-1G
103802-478EA 5 USD
103802-478 77109-626 77109-628 77109-630 77109-632 77298-914 77298-916 77315-450
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: >230 °C
Flash Pt: 166.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR