4-Hydroxynicotinic acid 97%

Supplier: Ambeed
Warning

A102032-100G A102032-10G A102032-250MG A102032-25G A102032-1G A102032-5G
103796-258EA 9 USD
103796-258 103821-956 76809-288 76809-290 76809-292 76809-294 77282-892 77308-842
4-Hydroxynicotinic acid 97%
4-Hydroxynicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 312.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00040286
CAS Number: 609-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR