2,4-Dicyanophenol 98%

Supplier: Ambeed
Warning

A114177-250MG A114177-1G
103798-356EA 142 USD
103798-356 103824-196 76971-076 77311-016
2,4-Dicyanophenol 98%
2,4-Dicyanophenol

Precaución: for Research Use Only

Formula: C₈H₄N₂O
Boiling Pt: 319.3 °C (760 mm Hg)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD11870755
CAS Number: 34133-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR