4-Hydroxyindanone 98%

Supplier: Ambeed
Warning

Synonyms: 4-HYDROXY-2,3-DIHYDRO-1H-INDEN-1-ONE, 4-hydroxyindan-1-one, 4-Hydroxy-1-indanone

A452561-25G A452561-5G A452561-100G A452561-10G A452561-500G A452561-1G
103827-804EA 9 USD
103827-804 103838-874 76842-706 76842-708 77116-210 77116-212 77116-214 77307-368
4-Hydroxyindanone 98%
4-Hydroxyindanone

Precaución: for Research Use Only

Formula: C₉H₈O₂
MW: 148.161 g/mol
Boiling Pt: 307.6 °C (760 mm Hg)
Melting Pt: 242…244 °C
Flash Pt: 130.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00143330
CAS Number: 40731-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR