4-Hydroxycyclohexanone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxycyclohexanon

A100030-1G A100030-5G A100030-100G A100030-10G A100030-250MG A100030-25G
103795-212EA 20 USD
103795-212 103820-934 77296-550 77296-552 77282-406 77282-408 77282-410 77282-412
4-Hydroxycyclohexanone 98%
4-Hydroxycyclohexanone

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.1424 g/mol
Boiling Pt: 226.3 °C (760 mm Hg)
Melting Pt: 1 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD02093817
CAS Number: 13482-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR