4-Hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A210882-100G A210882-500G A210882-1000G A210882-25G
103809-458EA 57 USD
103809-458 76873-670 76873-672 77303-154 77303-156
4-Hydroxybenzonitrile 98%
4-Hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅NO
Melting Pt: 110…113 °C
Flash Pt: 123.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00002312
CAS Number: 767-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR