1,5,6,7-Tetrahydro-4-hydroxy-2H-cyclopenta[b]pyridin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-6,7-dihydro-1H-cyclopenta[b]pyridin-2(5H)-one

A329140-100MG A329140-250MG A329140-5G
103825-516EA 99 USD
103825-516 103836-470 77194-970 77194-972 77306-392
1,5,6,7-Tetrahydro-4-hydroxy-2H-cyclopenta[b]pyridin-2-one 97%
1,5,6,7-Tetrahydro-4-hydroxy-2H-cyclopenta[b]pyridin-2-one

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Boiling Pt: 324.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09908208
CAS Number: 55618-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR