5-Methylpyrimidin-4-ol 95%

Supplier: Ambeed
Warning

A273168-100MG A273168-10G A273168-250MG A273168-25G A273168-5G
103813-132EA 17 USD
103813-132 103853-048 76772-178 76772-180 76772-182 77092-842 77092-844
5-Methylpyrimidin-4-ol 95%
5-Methylpyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
Boiling Pt: 192.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01646066
CAS Number: 17758-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR