4-Hydroxy-5,6-dimethylpyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A101696-250MG A101696-1G
103796-096EA 213 USD
103796-096 76982-190 77282-808
4-Hydroxy-5,6-dimethylpyridin-2(1H)-one 95%
4-Hydroxy-5,6-dimethylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD11113396
CAS Number: 84953-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR