4-(4-Hydroxyphenyl)benzonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 4'-Hydroxy-4-biphenylcarbonitrile, 4'-Hydroxybiphenyl-4-carbonitrile, 4'-Hydroxy-[1,1'-biphenyl]-4-carbonitrile

A200149-1G A200149-5G A200149-100G A200149-10G A200149-500G A200149-25G
103817-168EA 5 USD
103817-168 103847-796 77085-852 77085-854 77112-146 77112-148 77112-150 77302-654
4-(4-Hydroxyphenyl)benzonitrile 95%
4-(4-Hydroxyphenyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 191…195 °C
Flash Pt: 183.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00059625
CAS Number: 19812-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR