4-Hydroxy-3-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A144164-10G A144164-5G
103841-442EA 10 USD
103841-442 77137-458 77316-682
4-Hydroxy-3-nitrobenzonitrile 98%
4-Hydroxy-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
Boiling Pt: 288.9 °C (760 mm Hg)
Melting Pt: 146…148 °C
Flash Pt: 128.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00070775
CAS Number: 3272-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR