4-Hydroxy-3-methylquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A105077-100MG A105077-250MG A105077-1G A105077-5G
103796-834EA 173 USD
103796-834 103822-560 77160-440 77160-442 77309-432 77309-434
4-Hydroxy-3-methylquinolin-2(1H)-one 97%
4-Hydroxy-3-methylquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
Boiling Pt: 309.2 °C (760 mm Hg)
Flash Pt: 140.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00100774
CAS Number: 1873-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR