4-Hydroxy-3-methylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-m-toluic acid

A316704-1G A316704-5G A316704-500G
103836-042EA 10 USD
103836-042 77251-072 77306-152 77746-607
4-Hydroxy-3-methylbenzoic acid 98%
4-Hydroxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 331.3 °C (760 mm Hg)
Melting Pt: 173…177 °C
Flash Pt: 168.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00270105
CAS Number: 499-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR