4-Methoxy-2-methylphenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3-methylanisol

A132101-100G A132101-10G A132101-250MG A132101-1G A132101-25G A132101-5G
103801-688EA 30 USD
103801-688 76811-256 76811-258 76811-260 77298-536 77298-538 77298-540
4-Methoxy-2-methylphenol 98%
4-Methoxy-2-methylphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11055138
CAS Number: 5307-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR