4-Hydroxy-3-iodo-5-nitrobenzonitrile 97%

Supplier: Ambeed
Danger

A941421-1G A941421-5G A941421-100G A941421-250MG
103841-430EA 8 USD
103841-430 76845-700 76845-702 77118-940 77118-942
4-Hydroxy-3-iodo-5-nitrobenzonitrile 97%
4-Hydroxy-3-iodo-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃IN₂O₃
Boiling Pt: 280.1 °C (760 mm Hg)
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00070776
CAS Number: 1689-89-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR