3,5-Dimethyl-4-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3,5-dimethylbenzonitrile

A531232-25G A531232-10G A531232-5G
103839-678EA 5 USD
103839-678 76843-694 77279-888 77307-740
3,5-Dimethyl-4-hydroxybenzonitrile 98%
3,5-Dimethyl-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 310.6 °C (760 mm Hg)
Melting Pt: 123…127 °C
Flash Pt: 141.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00221716
CAS Number: 4198-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR