Syringaldehyde 98%

Supplier: Ambeed
Warning

A106877-1000G A106877-100G A106877-25G A106877-5G A106877-500G
103822-850EA 9 USD
103822-850 77245-476 77245-478 77245-480 77309-712 77570-414
Syringaldehyde 98%
Syringaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
Boiling Pt: 192…193 °C (760 mm Hg)
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006943
CAS Number: 134-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR