4-Hydroxy-3-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A292980-1G A292980-25G A292980-5G A292980-10G A292980-250MG
103813-962EA 19 USD
103813-962 103835-102 76837-712 76837-714 76837-716 77289-830 77289-832
4-Hydroxy-3-(trifluoromethyl)benzonitrile 98%
4-Hydroxy-3-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃NO
Boiling Pt: 256 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06797882
CAS Number: 124811-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR