4-Hydroxy-2-piperidinone 97%

Supplier: Ambeed
Warning

A133644-250MG A133644-1G A133644-5G
103801-892EA 27 USD
103801-892 77109-396 77314-564 77314-566
4-Hydroxy-2-piperidinone 97%
4-Hydroxy-2-piperidinone

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.1305 g/mol
Boiling Pt: 352.3 °C (760 mm Hg)
Flash Pt: 166.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09743648
CAS Number: 476014-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR