4-Amino-3-nitrophenol 98%

Supplier: Ambeed
Warning

A134828-100G A134828-10G A134828-500G A134828-25G A134828-5G
76811-458EA 37 USD
76811-458 76811-460 76811-462 77298-732 77298-734
4-Amino-3-nitrophenol 98%
4-Amino-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
Boiling Pt: 374.4 °C (760 mm Hg)
Melting Pt: 150…154 °C
Flash Pt: 180.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00066310
CAS Number: 610-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR