4-Hydroxy-2-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpyridin-4-ol

A219973-1G A219973-25G A219973-5G A219973-100G A219973-10G A219973-250MG
103818-830EA 6 USD
103818-830 103849-712 77088-294 77088-296 77088-298 77288-184 77288-186 77288-188
4-Hydroxy-2-methylpyridine 97%
4-Hydroxy-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.1259 g/mol
Boiling Pt: 201.2 °C (760 mm Hg)
Flash Pt: 98.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04114156
CAS Number: 18615-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR