4-Hydroxy-2-methylphenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
A321671-250MG
A321671-5G
A321671-1G
A321671-25G
103825-288EA
29
USD
103825-288
103836-220
76772-928
76839-132
77306-246
77306-248
4-Hydroxy-2-methylphenylboronic acid 98%
4-Hydroxy-2-methylphenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₉BO₃ Boiling Pt: 361.4 °C (760 mm Hg) Melting Pt: 100…104 °C Flash Pt: 172.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788424 CAS Number: 493035-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...