4-Hydroxy-2,6-dimethylbenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethyl-4-hydroxybenzaldehyde

A112168-100G A112168-10G A112168-25G A112168-500G A112168-1G A112168-5G
103797-968EA 5 USD
103797-968 103823-804 76809-982 76809-984 76809-986 76809-988 77284-214 77310-614
4-Hydroxy-2,6-dimethylbenzaldehyde 97%
4-Hydroxy-2,6-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.1700 g/mol
Boiling Pt: 283.8 °C (760 mm Hg)
Melting Pt: 190…196 °C
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016979
CAS Number: 70547-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR