4-Hydrazinylpyridine hydrochloride 95%

Supplier: Ambeed
Warning

A115132-250MG A115132-1G A115132-25G
103798-520EA 6 USD
103798-520 103824-364 77284-460 77311-184 77570-886
4-Hydrazinylpyridine hydrochloride 95%
4-Hydrazinylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈ClN₃
MW: 145.5900 g/mol
Boiling Pt: 310.2 °C (760 mm Hg)
Flash Pt: 141.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035353
CAS Number: 20815-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR